Learn More
Aviva Systems Biology Corp CST9 Antibody - middle region (ARP59193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CST9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CST9. Alternate names: CLMApplicationWB. Species reactivity: Human
Catalog No. 50-286-615