Learn More
Aviva Systems Biology Corp CSTB Antibody - N-terminal region (ARP45749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSTB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CSTB. Alternate names: CST6, EPM1, PME, STFB, ULD, EPM1AApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-59127