Learn More
Aviva Systems Biology Corp CSTF2T Antibody - C-terminal region (ARP40808_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSTF2T. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CSTF2T. Alternate names: CstF-64T, DKFZp434C1013, KIAA0689ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57628