Learn More
Aviva Systems Biology Corp CSTF3 Antibody - middle region (ARP40353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSTF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSTF3. Alternate names: CSTF-77, MGC117398, MGC43001, MGC75122ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57406