Learn More
Electron Microscopy Sciences Aurion ImmunoGold Custom Ultra Small EM Kit
SDP
Catalog No. 5028290
Encompass_Preferred
Immunogold reagent kit. Two non-overlapping gold particle sizes. Coupled to: either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies. For customization choose two vials of Ultra-Small Reagent and 30ml BSA-c and 30ml SE-EM.
Catalog No. 50-282-90 Supplier Electron Microscopy Sciences Supplier No. 2555006
Immunogold reagent kit. Two non-overlapping gold particle sizes. Coupled to: either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies. For customization choose two vials of Ultra-Small Reagent and 30ml BSA-c and 30ml SE-EM.