Learn More
Leica Biosystems CSTM WHITE MB CASSETTES 1000/C
Supplier:  Leica Biosystems 3802731
Encompass_Preferred
Microbiopsy Cassettes are specially designed to accommodate extremely small specimens and tissue biopsies.These cassettes features a 0.3 mm pore size and have a separate compartment with larger holes for rapid fluid exchange to reduce flotation.The cassettes can easily be opened and closed with one hand.Available in seven colors in quantities of 250 or 1000.
Catalog No. 50-255-2430