Learn More
Medchemexpress LLC HY-P7150 10ug Medchemexpress, CT-1 (HEK293-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 502083115
Encompass_Preferred
Medchemexpress, HY-P7150 10ug CT-1 (HEK293-expressed), Mouse CAS: Recombinant Mouse Cardiotrophin-1 (HEK293-expressed), a member of IL-6 family of cytokines, is required for cardiac myocyte maturation and plays an important role in the differentiation, development, and survival of neural stem cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3115 Supplier Medchemexpress LLC Supplier No. HYP715010UG
Medchemexpress, HY-P7150 10ug CT-1 (HEK293-expressed), Mouse CAS: Recombinant Mouse Cardiotrophin-1 (HEK293-expressed), a member of IL-6 family of cytokines, is required for cardiac myocyte maturation and plays an important role in the differentiation, development, and survival of neural stem cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.