Learn More
Medchemexpress LLC CT-2584 | 166981-13-1 | 99.4% | 533.79 g/mol | C30H55N5O3 | 1 MG
SDP
Catalog No. 5000229880
Encompass_Preferred
CT-2584 (CAS 166981-13-1) is a small-molecule anticancer research compound reported to reduce NKEF-B expression and to kill tumor cells by inducing mitochondrial reactive oxygen species (ROS). It has a molecular formula of C30H55N5O3, a molecular weight of 533.79 g/mol, and is provided at high purity for laboratory research.
  • High reported purity (99.4%) suitable for research applications.
  • Mechanism: reduces NKEF-B expression and induces mitochondrial ROS in tumor cells.
  • Characterized molecular properties: formula C30H55N5O3, molar mass 533.79 g/mol.
  • Typically supplied in small research quantities for in vitro studies.
Catalog No. 50-002-29880 Supplier Medchemexpress LLC Supplier No. HY1189821MG
CT-2584 (CAS 166981-13-1) is a small-molecule anticancer research compound reported to reduce NKEF-B expression and to kill tumor cells by inducing mitochondrial reactive oxygen species (ROS). It has a molecular formula of C30H55N5O3, a molecular weight of 533.79 g/mol, and is provided at high purity for laboratory research.
  • High reported purity (99.4%) suitable for research applications.
  • Mechanism: reduces NKEF-B expression and induces mitochondrial ROS in tumor cells.
  • Characterized molecular properties: formula C30H55N5O3, molar mass 533.79 g/mol.
  • Typically supplied in small research quantities for in vitro studies.