Learn More
Zymo Research Corporation CT Conversion Reagent (96 Conversions), 1 bottle
SDP
Supplier:  Zymo Research Corporation D50031
Encompass_Preferred
Bisulfite conversion reagent used to process up to 96 samples for methylation analysis. Used in the EZ DNA Methylation, EZ DNA Methylation - Gold, and EZ DNA Methylation - Direct kits.
Catalog No. 50-203-7106