Learn More
Perkin Elmer US LLC CT-Flex Corrugated Teflon FEP Tubing - 0.25 in OD
Catalog No. 502016476
Encompass_Preferred
CT-Flex Corrugated Teflon FEP Tubing shapes and flexes easily and can take tortuous paths. It features a practically zero-bend radius and can be extended or compressed without affecting the inner diameter. CT-Flex resists chemicals, elevated temperatures, cut-through, and strain. CT-Flex is made from Teflon FEP, has read-thru transparency and has FDA approval under regulation 21 CFR 17.1650. The non-wetting surface of CT-Flex means easy clean-out, so batch-to-batch contamination is not a problem. Teflon FEP contains no plasticizers or filler to leach out and contaminate the product stream. Other features include: Vacuum Service: 29.9 in Hg. Extension/Compression Length Ratio: 2:1 Max Continuous Service Temperature: 200°F (0 pressure)
Catalog No. 50-201-6476 Supplier Perkin Elmer US LLC Supplier No. 09908689
CT-Flex Corrugated Teflon FEP Tubing shapes and flexes easily and can take tortuous paths. It features a practically zero-bend radius and can be extended or compressed without affecting the inner diameter. CT-Flex resists chemicals, elevated temperatures, cut-through, and strain. CT-Flex is made from Teflon FEP, has read-thru transparency and has FDA approval under regulation 21 CFR 17.1650. The non-wetting surface of CT-Flex means easy clean-out, so batch-to-batch contamination is not a problem. Teflon FEP contains no plasticizers or filler to leach out and contaminate the product stream. Other features include: Vacuum Service: 29.9 in Hg. Extension/Compression Length Ratio: 2:1 Max Continuous Service Temperature: 200°F (0 pressure)