Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ CT/GC/TV Control Panel (Inactivated Pellet)
SDP

Catalog No. 23004143 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-143 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-143 Supplier Microbiologics™ Supplier No. 8228
Add to Cart
Add to Cart
This item is not returnable. View return policy

CT/GC/TV Control Control Panel (Inactivated Pellet) contains six inactivated, pre-pooled pellets comprised of microorganisms.

Each pellet contains the microorganisms: Chlamydia trachomatis, Neisseria gonorrhoeae, and Trichomonas vaginalis

Specifications

Biosafety Level 1
Content And Storage 2 to 25°C
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) Clinical
Packaging Type Plastic Box
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.