Learn More
Bellco Glass, Inc. CT II Digital.Over Head Drive Complete, 115V
SDP
Catalog No. 501946217
Encompass_Preferred
The Bellco Digital CTII Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixing and the stirring of solutions with a viscosity greater than water. The Overhead Drives feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Units are powered by a 24VDC power supply (included) and are CE compliant. A detachable speed cable (included) attaches to the rear of the Digital CTII to allow real-time monitoring and trending of data when connected to a Celltrol II controller.. MADE IN THE USA
Catalog No. 50-194-6217 Supplier Bellco Glass, Inc. Supplier No. 777450115
The Bellco Digital CTII Overhead Drive is a versatile direct-drive agitator for cell culture and most aseptic mixing applications. The Overhead Drives were developed to eliminate the problem of magnetic agitator decoupling often observed in large volume mixing and the stirring of solutions with a viscosity greater than water. The Overhead Drives feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Units are powered by a 24VDC power supply (included) and are CE compliant. A detachable speed cable (included) attaches to the rear of the Digital CTII to allow real-time monitoring and trending of data when connected to a Celltrol II controller.. MADE IN THE USA