Learn More
Grainger Marathon Electric AC Motor, 15 HP, 460V, TEFC, 256T Frame, 1 ea
Catalog No. 502792861
Encompass_Preferred
This industrial duty Globetrotter series motor is designed for continuous operation in cooling tower applications. It features a totally enclosed fan-cooled (TEFC) enclosure with cast iron construction and IP65 housing rating, providing robust protection in demanding environments.
  • 15 hp (high speed) and 3 3/4 hp (low speed) power ratings
  • 460 V AC operation at 60 Hz
  • NEMA 256T frame with rigid base
  • Class F insulation
  • Forsheda seals on both ends
  • Extended grease tubes and shaft slinger
  • T-drains provided for moisture management
  • Actual test and vibration data supplied with each unit
Catalog No. 50-279-2861 Supplier Grainger Supplier No. 19YR30
This industrial duty Globetrotter series motor is designed for continuous operation in cooling tower applications. It features a totally enclosed fan-cooled (TEFC) enclosure with cast iron construction and IP65 housing rating, providing robust protection in demanding environments.
  • 15 hp (high speed) and 3 3/4 hp (low speed) power ratings
  • 460 V AC operation at 60 Hz
  • NEMA 256T frame with rigid base
  • Class F insulation
  • Forsheda seals on both ends
  • Extended grease tubes and shaft slinger
  • T-drains provided for moisture management
  • Actual test and vibration data supplied with each unit