Learn More
Grainger Marathon Motors Cooling Tower Motor, 5 hp, 1,755 RPM, TEFC, Cast Iron, 1 ea
Catalog No. 502793012
Encompass_Preferred
This three-phase cooling tower motor is a robust, rigid-base unit designed for reliable operation in demanding environments. Featuring a totally enclosed fan-cooled (TEFC) design, this motor provides high starting torque and operational efficiency. It is suitable for both horizontal and vertical shaft-up mounting configurations, making it a versatile choice for machinery and cooling equipment applications.
  • 5 hp output at 1,755 RPM
  • Three-phase 230/460V AC power supply
  • Durable cast iron frame with epoxy-coated finish
  • IP55 rated for protection against dust and water jets
  • Inverter duty rated for 10:1 variable torque and 10:1 constant torque
  • Class F insulation for reliable thermal management
  • Ball bearings with 1.15 service factor
Catalog No. 50-279-3012 Supplier Grainger Supplier No. 19YR40
This three-phase cooling tower motor is a robust, rigid-base unit designed for reliable operation in demanding environments. Featuring a totally enclosed fan-cooled (TEFC) design, this motor provides high starting torque and operational efficiency. It is suitable for both horizontal and vertical shaft-up mounting configurations, making it a versatile choice for machinery and cooling equipment applications.
  • 5 hp output at 1,755 RPM
  • Three-phase 230/460V AC power supply
  • Durable cast iron frame with epoxy-coated finish
  • IP55 rated for protection against dust and water jets
  • Inverter duty rated for 10:1 variable torque and 10:1 constant torque
  • Class F insulation for reliable thermal management
  • Ball bearings with 1.15 service factor