Learn More
Grainger Marathon Cooling Tower Motor, TEFC, Cast Iron, 60/15 hp, 1,780/885 RPM, 1 ea
Catalog No. 502792673
Encompass_Preferred
This three-phase, two-speed cooling tower motor is designed for severe duty applications, providing high-efficiency performance and reliable, continuous operation. Engineered with a robust cast iron frame and epoxy-coated finish, this motor offers exceptional rigidity and vibration resistance in demanding industrial environments.
  • Blue Chip® quality construction
  • 100% cast iron housing
  • Two-speed, one-winding design
  • Class F insulation with 1.15 service factor
  • Regreasable bearings with extended grease tubes
  • Forsheda seals and shaft slinger on both ends for moisture protection
  • UL Recognized and CSA Certified
  • Includes T-drains for effective condensation management
  • Three-year warranty
Catalog No. 50-279-2673 Supplier Grainger Supplier No. 19YR36
Add to Cart
Add to Cart
Request a Quote
This three-phase, two-speed cooling tower motor is designed for severe duty applications, providing high-efficiency performance and reliable, continuous operation. Engineered with a robust cast iron frame and epoxy-coated finish, this motor offers exceptional rigidity and vibration resistance in demanding industrial environments.
  • Blue Chip® quality construction
  • 100% cast iron housing
  • Two-speed, one-winding design
  • Class F insulation with 1.15 service factor
  • Regreasable bearings with extended grease tubes
  • Forsheda seals and shaft slinger on both ends for moisture protection
  • UL Recognized and CSA Certified
  • Includes T-drains for effective condensation management
  • Three-year warranty