Learn More
Perkin Elmer US LLC CT Q+ MEINHARD Plus Quartz Nebulizer for NexION 1000/2000
Catalog No. 502112170
Encompass_Preferred
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer. This CT Q+ MEINHARD Plus Nebulizer is included in the Organics Sample Introduction Kit, Orange, for NexION 1000/2000 (N8150031).
Catalog No. 50-211-2170 Supplier Perkin Elmer US LLC Supplier No. N8152372
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer. This CT Q+ MEINHARD Plus Nebulizer is included in the Organics Sample Introduction Kit, Orange, for NexION 1000/2000 (N8150031).