Learn More
Perkin Elmer US LLC CT R+ MEINHARD Plus Glass Nebulizer for NexION 1000/2000
Catalog No. 502112171
Encompass_Preferred
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer. This CT R+ MEINHARD Plus Nebulizer is included in the Sample Introduction Kit for NexION 1000 - Purple (N8150038), Sample Introduction Kit for NexION 2000 Base Configuration - Blue (N8150034), and the Sample Introduction Kit for NexION 2000 Core Configuration - Green (N8150035).
Catalog No. 50-211-2171 Supplier Perkin Elmer US LLC Supplier No. N8152373
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer. This CT R+ MEINHARD Plus Nebulizer is included in the Sample Introduction Kit for NexION 1000 - Purple (N8150038), Sample Introduction Kit for NexION 2000 Base Configuration - Blue (N8150034), and the Sample Introduction Kit for NexION 2000 Core Configuration - Green (N8150035).