Learn More
Sino Biological Recombinant Human EPOR & CD131 Heterodimer Protein (hFc Tag)
Supplier:  Sino Biological CT010H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR (NP_000112.1) linked with the extracellular domain (Trp 17-Trp 443) of human CD131 (NP_000386.1) by a peptide linker was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-202-2313