Learn More
Sino Biological Recombinant Human CD3D & CD3E Heterodimer Protein (ECD, human IgG1-Fc & His & human IgG1-Fc & Flag Tag), HPLC-verified
Supplier:  Sino Biological CT026H0323H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ala105) of human CD3D (P04234-1) was fused with the C-terminal his-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asp126) of human CD3E (P07766-1) was fused with the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD3D/CD3E heterodimer was purified.
Catalog No. 50-202-2345