Learn More
Sino Biological Human CD3D & CD3E Heterodimer Protein (Flag & His Tag) 50ug
Supplier:  Sino Biological CT038H2508H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ala105) of human CD3D (P04234) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asp126) of human CD3E (P07766) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human CD3D/CD3E heterodimer was purified.
Catalog No. 50-202-2369