Learn More
Sino Biological Recombinant Human IL-12 Protein, HPLC-verified
Supplier:  Sino Biological CT050HNAH10
Encompass_Preferred
A DNA sequence encoding the human IL12A (P29459-1)(Met1-Ser219) was expressed, constructed the plasmid 1; A DNA sequence encoding the human IL12B (NP_002178.2)(Met1-Ser328) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the human IL12A&IL12B heterodimer was purified.
Catalog No. 50-202-2401