Learn More
ATCC CT26.WT; Colon Carcinoma; Mouse (Mus musculus)
Catalog No. 502383144
Encompass
CT26 is an N-nitroso-N-methylurethane-(NNMU) induced, undifferentiated colon carcinoma cell line. It was cloned to generate the cell line designated CT26.WT (ATCC CRL-2638). Use CT26.WT mouse fibroblasts with CT26.CL25 (ATCC CRL-2639) as a model for testing immunotherapy protocols and in studies on the host immune response.
Catalog No. 50-238-3144 Supplier ATCC Supplier No. CRL2638
CT26 is an N-nitroso-N-methylurethane-(NNMU) induced, undifferentiated colon carcinoma cell line. It was cloned to generate the cell line designated CT26.WT (ATCC CRL-2638). Use CT26.WT mouse fibroblasts with CT26.CL25 (ATCC CRL-2639) as a model for testing immunotherapy protocols and in studies on the host immune response.