Learn More
Medchemexpress LLC CT83 Human His B2 10ug
SDP
Catalog No. 5000196378
Encompass_Preferred
Recombinant human CT83 protein supplied as a lyophilized preparation with N-terminal His and B2M tags. The protein is expressed in Escherichia coli, exhibits >90% purity by reducing SDS-PAGE, and is provided in a 10 μg quantity. It is formulated in Tris-EDTA with trehalose for stability and is suitable as a reagent for immunology, cancer antigen research, antibody development, and assay controls.
  • Recombinant human CT83 protein with N-His and B2M tags.
  • Expressed in Escherichia coli and lyophilized for shelf stability.
  • Greater than 90% purity as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 26.8 kDa.
  • Supplied as a 10 μg quantity, suitable for assay development.
  • Lyophilized formulation contains Tris-HCl, EDTA, and 6% trehalose.
  • Store frozen and aliquot for long-term stability after reconstitution.
Catalog No. 50-001-96378 Supplier Medchemexpress LLC Supplier No. HYP7156210UG
Recombinant human CT83 protein supplied as a lyophilized preparation with N-terminal His and B2M tags. The protein is expressed in Escherichia coli, exhibits >90% purity by reducing SDS-PAGE, and is provided in a 10 μg quantity. It is formulated in Tris-EDTA with trehalose for stability and is suitable as a reagent for immunology, cancer antigen research, antibody development, and assay controls.
  • Recombinant human CT83 protein with N-His and B2M tags.
  • Expressed in Escherichia coli and lyophilized for shelf stability.
  • Greater than 90% purity as determined by reducing SDS-PAGE.
  • Approximate molecular weight of 26.8 kDa.
  • Supplied as a 10 μg quantity, suitable for assay development.
  • Lyophilized formulation contains Tris-HCl, EDTA, and 6% trehalose.
  • Store frozen and aliquot for long-term stability after reconstitution.