Learn More
Aviva Systems Biology Corp CTA-126B4.3 Antibody - N-terminal region (ARP40817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTA-126B4.3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CTA-126B4.3. Alternate names: BK126B4.3, CGI-96, MGC150422, MGC150423, CTA-126B4.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53241