Learn More
Bioworld CTAB, 100 g
Catalog No. 50255819
Encompass_Preferred
Cetyltrimethylammonium bromide A cationic detergent used in the isolation of high molecular weight DNAAlso known as CTAB (citrimonium bromide)High Purity GradeUsed to precipitate DNAAppearance: White powderHS Code : 29239000IUPAC_Name: Hexadecyl-Trimethyl-Ammonium BromideMelting Point: 248-251 °C (lit.)RTECS Number: BQ7875000HS Code: 2923.90.00
Catalog No. 50-255-819 Supplier Bioworld Supplier No. 403300443
Cetyltrimethylammonium bromide A cationic detergent used in the isolation of high molecular weight DNAAlso known as CTAB (citrimonium bromide)High Purity GradeUsed to precipitate DNAAppearance: White powderHS Code : 29239000IUPAC_Name: Hexadecyl-Trimethyl-Ammonium BromideMelting Point: 248-251 °C (lit.)RTECS Number: BQ7875000HS Code: 2923.90.00