Learn More
Teknova CTAB Extraction Solution. 250mL. 4 Pack.
SDP
Catalog No. 502387315
Encompass
2.0% CTAB, 100mM Tris-HCl, pH 8.0,20mM EDTA, pH 8.0, 1.4M Sodium Chloride
Catalog No. 50-238-7315 Supplier Teknova Supplier No. C21904PK
2.0% CTAB, 100mM Tris-HCl, pH 8.0,20mM EDTA, pH 8.0, 1.4M Sodium Chloride