Learn More
Aviva Systems Biology Corp CTBS Antibody - C-terminal region (ARP63779_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63779_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTBS. It was validated on Western Blot. . Alternate names: CTBApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-081