Learn More
Rarecyte RareCyte RarePlex CTC Negative Control Slide, 10-Pack
Catalog No. NC1854148
Encompass
Batch control slides to be stained, scanned, and reviewed alongside experimental slides. Slides contain a monolayer of buffy coat from healthy normal donors. Slides are negative for circulating tumor cells (CTCs).
Catalog No. NC1854148 Supplier Rarecyte Supplier No. 241278000
Batch control slides to be stained, scanned, and reviewed alongside experimental slides. Slides contain a monolayer of buffy coat from healthy normal donors. Slides are negative for circulating tumor cells (CTCs).