Learn More
Aviva Systems Biology Corp CTCFL Antibody - N-terminal region (ARP36128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTCFL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CTCFL. Alternate names: BORIS, CTCF-T, MGC163358, dJ579F20.2, CT27, HMGB1L1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53031