Learn More
Seracare Life Sciences Inc ctDNA Mutation Mix v2 AF0.25%, 25 uL
Catalog No. NC1814091
Encompass
To overcome the lack of patient-like reference materials for liquid biopsy, including drawbacks of existing methodologies such as sonication, SeraCare has developed a breakthrough technology that produces the most patient-like materials compared to native ctDNA. The Seraseq ctDNA Mutation Mix v2 is a highly multiplexed product for NGS-based ctDNA assays targeting cancer-relevant somatic mutations.
This item is not returnable. View return policy
Catalog No. NC1814091 Supplier Seracare Life Sciences Inc Supplier No. 07100142
To overcome the lack of patient-like reference materials for liquid biopsy, including drawbacks of existing methodologies such as sonication, SeraCare has developed a breakthrough technology that produces the most patient-like materials compared to native ctDNA. The Seraseq ctDNA Mutation Mix v2 is a highly multiplexed product for NGS-based ctDNA assays targeting cancer-relevant somatic mutations.