Learn More
Aviva Systems Biology Corp CTDSPL Antibody - N-terminal region (ARP42261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTDSPL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CTDSPL. Alternate names: C3orf8, HYA22, PSR1, SCP3, RBSP3ApplicationIHC, WB. Species reactivity: Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-53332