Learn More
Selleck Chemical LLC CTEP (RO4956371) S2861-5mg
SDP
Catalog No. 503112332
Encompass_Preferred
CTEP (RO4956371) is a novel long-acting orally bioavailable allosteric antagonist of mGlu5 receptor with IC50 of 2 2 nM shows 1000-fold selectivity over other mGlu receptors
Catalog No. 50-311-2332 Supplier Selleck Chemical LLC Supplier No. S28615MG
CTEP (RO4956371) is a novel long-acting orally bioavailable allosteric antagonist of mGlu5 receptor with IC50 of 2 2 nM shows 1000-fold selectivity over other mGlu receptors