Learn More
Aviva Systems Biology Corp CTF1 Antibody - N-terminal region (ARP51944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CTF1. Alternate names: CT-1, CT1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-612