Learn More
Perkin Elmer US LLC CTFE Stator for HTS Valve
Catalog No. 502117281
Encompass_Preferred
PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. This vacuum-based, valve-driven system is fully integrated with SyngistixTM 2.5 Software, eliminating the need for third party software and reducing troubleshooting. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.
Catalog No. 50-211-7281 Supplier Perkin Elmer US LLC Supplier No. N8152938
PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. This vacuum-based, valve-driven system is fully integrated with SyngistixTM 2.5 Software, eliminating the need for third party software and reducing troubleshooting. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.