Learn More
SOLVENTUM 3M™ LifeASSURE™ PFS Series Filter Cartridge
Supplier:  SOLVENTUM 7000029190
Encompass_Preferred
This item has an order quantity of multiples of 6 per supplier requirements. Flow-enhancing construction allows air flow with a relatively low pressure drop, resulting in a smaller vent filter assemblies. Hydrophobic polytetrafluoroethylene (PTFE) membrane helps prevent pore blockage in vent applications, extending filter lifespan. Liquid validation of B. diminuta retention helps provide reliable sterilizing performance in wet or dry conditions
Catalog No. NC0834075