Learn More
Enzo Life Sciences Connective tissue growth factor (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2120100
Encompass_Preferred
Alternative name: CCN2, HCS24, CTGF. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized. Contains 10 mM sodium acetate, pH 6.0. Source: Produced in E. coli. A non-glycosylated protein containing 98 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4002