Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP537320UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Human. BioActivity: Bioactivity of Human CTGF confirmed in HUVEC cell proliferation assay. AA Sequence: . MGKKCIRTP K. ISKPIKFE LS. GCTSMKT YRA. KFCGVC TDGR. CCTPH RTTTL. PVEF KCPDGE. VMK KNMMFIK. TC ACHYNCPG. D NDIFESLYY.  RKMYGDMA. Buffer: Sterile filtered (0.22 um) solution containing 0.1% TFA. Purity: Greater than 90 as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: CTGF. Gene ID: 1490. Uniprot: P29279.
Catalog No. 50-188-5800