Learn More
ACROBiosystems Recombinant Protein;Mouse CTGF / CCN2 Protein;HEK293;1MG;CTF,CCN2,NOV2,HCS24,IGFBP8,CTGF,IBP-8,IGFBP-8,IGF-binding protein 8
Supplier:  ACROBiosystems CTFM52HC1MG
Encompass_Preferred
Recombinant Protein;Mouse CTGF / CCN2 Protein;1MG;Mouse CTGF (CTF-M52Hc) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Ala 348 (Accession # P29268-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.3 kDa. The protein migrates as 37-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7622