Learn More
ACROBiosystems Recombinant Protein;Human CTGF / CCN2 Protein;HEK293;1MG;CTF,CCN2,NOV2,HCS24,IGFBP8,CTGF,IBP-8,IGFBP-8,IGF-binding protein 8
Catalog No. 501657572
Encompass_Preferred
Recombinant Protein;Human CTGF / CCN2 Protein;1MG;Human CTGF (CTF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # Q5M8T4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.4 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7572 Supplier ACROBiosystems Supplier No. CTFH52H51MG
Recombinant Protein;Human CTGF / CCN2 Protein;1MG;Human CTGF (CTF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession # Q5M8T4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.4 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.