Learn More
FUJIFILM Biosciences CTGrade GMP rh PDGF-AA
Catalog No. 502892147
Encompass_Preferred
Platelet-derived growth factor (PDGF) serves as a crucial controller of cell growth, proliferation, and angiogenesis (1). PDGF exists as a dimeric glycoprotein, comprising two polypeptide chains (A and B) stabilized by disulfide bridges. These chains can combine to form either heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (2) PDGF-AA is commonly used for differentiation of neural progenitor cells derived from human pluripotent stem cells (hPSC) into oligodendrocyte precursor cells (3). The synthesis of PDGF is stimulated by signaling molecules such as IL-1, IL-6, TNF-A, TGF-B, and EGF. PDGF acts as a growth hormone that induces cell division in mesenchymal lineage cells like smooth muscle and glial cells. Furthermore, PDGF is stored within the alpha-granules of platelets and is released when these platelets adhere to injured tissues. The PDGF dimer binds to cell surface receptor tyrosine kinases, specifically PDGFR-A and PDGFR-B (4).
Catalog No. 50-289-2147 Supplier FUJIFILM Biosciences Supplier No. 5002350UG
Platelet-derived growth factor (PDGF) serves as a crucial controller of cell growth, proliferation, and angiogenesis (1). PDGF exists as a dimeric glycoprotein, comprising two polypeptide chains (A and B) stabilized by disulfide bridges. These chains can combine to form either heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (2) PDGF-AA is commonly used for differentiation of neural progenitor cells derived from human pluripotent stem cells (hPSC) into oligodendrocyte precursor cells (3). The synthesis of PDGF is stimulated by signaling molecules such as IL-1, IL-6, TNF-A, TGF-B, and EGF. PDGF acts as a growth hormone that induces cell division in mesenchymal lineage cells like smooth muscle and glial cells. Furthermore, PDGF is stored within the alpha-granules of platelets and is released when these platelets adhere to injured tissues. The PDGF dimer binds to cell surface receptor tyrosine kinases, specifically PDGFR-A and PDGFR-B (4).