Learn More
Aviva Systems Biology Corp CTH Antibody - C-terminal region (ARP46068_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46068_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CTH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CTH. Alternate names: MGC9471ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59206