Learn More
CTI Polypropylene Shoe Covers
GREENER_CHOICE
Catalog No. 191305576
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19-130-5576 X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-130-5576 Supplier CTI Supplier No. SCR645XL150
Add to Cart
Add to Cart
This item is not returnable. View return policy
Strong and durable shoe covers are heavilty textured for a sure grip and are compatible with class 100 environments.
  • Lightweight material offers comfort and breathability, while maintaing snug, full coverage
  • Individually inspected for stitching, sizing, materials and workmanship
  • Lot traceability
  • Color: Blue
Specifications
Size X-Large
Material Polypropylene
Color Blue
Sole Type Sewn Bottom
Thread Color Black
Product Type Controlled Environments Shoe Covers