Learn More
CELLULAR TECHNOLOGY LTD Ctl Anti-Aggregate Wash 20X, Size 1 ml
SDP
Supplier:  CELLULAR TECHNOLOGY LTD CTLAA001
Encompass
Ctl Anti-Aggregate Wash 20X, Size 1 Ml. Stabilizes Pbmc During Thawing And Prevents Cell Loss Through Aggregation. Cells That Die During Freeze-Thawing Release Dna Strands That Can Cause Cell Clumping And Aggregation. Therefore, The Use Of Dnase-Containing Wash Solutions Is Recommended While Thawing Pbmc. Moreover, Cells Show Increased Metabolic Activity After Thawing As They Compensate For The Stress Reaction - This Needs To Be Accounted For By Using A Nutrient-Rich Medium. Ctl Anti-Aggregate Wash Supplement Has Been Specifically Formulated For Optimal Pbmc Recovery And Functionality After Thawing. It Contains Injection-Grade Water Supplemented With Amino Acids, Vitamins, Inorganic Salts, And Human Serum Albumin (Hsa) In A Proprietary Composition, Saturated And Unsaturated Fatty Acids, And Sugars In A Ready-To-Use Formulation, Providing A Balanced Environment For Pbmc. For Each Vial Of Pbmc To Be Thawed, 1 Vial Of Ctl Anti-Aggregate Wash Supplement 20X Is Needed.
Catalog No. NC0089914