Learn More
CELLULAR TECHNOLOGY LTD CTL TEST MEDIA 500ML
SDP
Catalog No. nc9711916
Encompass
CTL-Test(TM) Medium CTL-Test(TM) Medium has been formulated to permit direct ex vivo testing of PBMC in T cell cytokine assays (ELISPOT, ELISA, CBA, CPA), while showing an equal or better performance than pre-screened, non-mitogenic serum batches. CTL-Test(TM) Medium contains injection grade water supplemented with amino acids, vitamins, inorganic salts, and human serum albumin (HSA) in a proprietary composition to provide a nutritionally complete and balanced environment for the PBMC. A variant of this medium, CTL-Test B(TM), has been specially formulated for B cell ELISPOT assays. CTL-Test(TM) Medium is available in 100ml or 500ml bottles. The 100ml bottles are convenient for average-sized experiments, avoiding the need for sterile filtration, saving the cost of filters. Each batch of CTL-Test(TM) Medium is thoroughly tested for an equally high performance in ELISPOT assays as the previous batches, in addition to standard quality assurance procedures.
Catalog No. NC9711916 Supplier CELLULAR TECHNOLOGY LTD Supplier No. CTLT005
CTL-Test(TM) Medium CTL-Test(TM) Medium has been formulated to permit direct ex vivo testing of PBMC in T cell cytokine assays (ELISPOT, ELISA, CBA, CPA), while showing an equal or better performance than pre-screened, non-mitogenic serum batches. CTL-Test(TM) Medium contains injection grade water supplemented with amino acids, vitamins, inorganic salts, and human serum albumin (HSA) in a proprietary composition to provide a nutritionally complete and balanced environment for the PBMC. A variant of this medium, CTL-Test B(TM), has been specially formulated for B cell ELISPOT assays. CTL-Test(TM) Medium is available in 100ml or 500ml bottles. The 100ml bottles are convenient for average-sized experiments, avoiding the need for sterile filtration, saving the cost of filters. Each batch of CTL-Test(TM) Medium is thoroughly tested for an equally high performance in ELISPOT assays as the previous batches, in addition to standard quality assurance procedures.