Learn More
CELLULAR TECHNOLOGY LTD CTL-Test Plus Medium
SDP
Catalog No. NC0837250
Encompass
CTL-Test PLUS Medium is an enhanced, buffered, serum-free solution containing injection-grade water supplemented with amino acids, vitamins, inorganic salts, and HSA in a proprietary composition to provide a nutritionally complete and balanced environment for PBMC.
Catalog No. NC0837250 Supplier CELLULAR TECHNOLOGY LTD Supplier No. CTLTP005
CTL-Test PLUS Medium is an enhanced, buffered, serum-free solution containing injection-grade water supplemented with amino acids, vitamins, inorganic salts, and HSA in a proprietary composition to provide a nutritionally complete and balanced environment for PBMC.