Learn More
AURORA BIOLABS LLC CTLA-4/B7-1 Inhibitor Binding Assay Kit,384 rxn
Catalog No. NC4007643
Encompass_Preferred
The CTLA-4/B7-1 binding assay kit is a TR-FRET based assay, which is designed to detect the binding status between CTLA-4 and B7-1. Tag6-CTLA-4 and Tag7-B7-1 are included in this assay kit. Binding of Tag6-CTLA-4 to Tag7-B7-1 brings the Terbium (Tb, HTRF donor) and the fluorophore d2 (HTRF acceptor) in a proximity distance, and activation of Tb results in fluorescence resonance energy transfer (FRET). Thus, the binding status can be quantitively measured by calculating the ratio of the emission fluorescence intensity of the acceptor (665 nm) and donor (620 nm). Interference of the CTLA-4/B7-1 binding will reduce the HTRF signal.
Catalog No. NC4007643 Supplier AURORA BIOLABS LLC Supplier No. 232852384
Add to Cart
Add to Cart
The CTLA-4/B7-1 binding assay kit is a TR-FRET based assay, which is designed to detect the binding status between CTLA-4 and B7-1. Tag6-CTLA-4 and Tag7-B7-1 are included in this assay kit. Binding of Tag6-CTLA-4 to Tag7-B7-1 brings the Terbium (Tb, HTRF donor) and the fluorophore d2 (HTRF acceptor) in a proximity distance, and activation of Tb results in fluorescence resonance energy transfer (FRET). Thus, the binding status can be quantitively measured by calculating the ratio of the emission fluorescence intensity of the acceptor (665 nm) and donor (620 nm). Interference of the CTLA-4/B7-1 binding will reduce the HTRF signal.