Learn More
BIO SB INC CTLA-4 - CD152 1ML
Supplier:  BIO SB INC BSB2885 (ASR)
Encompass
CTLA-4 antibody (CD152 antibody) is a mouse monoclonal antibody derived from cell culture supernatant that is concentrated, dialyzed, filter sterilized and diluted in buffer pH 7.5, containing BSA and sodium azide as a preservative. 1 ML. Clone: BSB-88. Isotype: IgG2a/K
Catalog No. NC2480558