Learn More
ACROBiosystems ACROBiosystems Rat CTLA-4 / CD152 Protein, Fc Tag
Supplier:  ACROBiosystems CT4R52591MG
Encompass_Preferred
Recombinant Protein;1MG;Rat CTLA-4, Fc Tag (CT4-R5259) is expressed from human 293 cells (HEK293). It contains AA Ile 38 - Asp 161 (Accession # Q62859).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse B7-2, His Tag (Cat. No. CD6-M52H0) at 2 μg/mL (100 μL/well) can bind Rat CTLA-4, Fc Tag (Cat. No. CT4-R5259) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-7330