Learn More
Medchemexpress LLC CTLA-4 Guinea pig 10ug
SDP
Supplier:  Medchemexpress LLC HYP7198610UG
Encompass_Preferred
Recombinant His-tagged extracellular domain of guinea pig CTLA-4 produced in Pichia pastoris and supplied as a sterile, 0.2 μm-filtered PBS solution for research use. The protein is >90% pure by reducing SDS-PAGE, has an approximate molecular weight of 16-35 kDa, and is provided in multiple microgram sizes for biochemical and binding studies. Store frozen and avoid repeated freeze-thaw cycles for best stability.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • C-terminal His tag for affinity purification and detection.
  • Produced in Pichia pastoris expression system.
  • Supplied in 0.2 μm-filtered PBS, pH 7.4, ready for assays.
  • Multiple sizes available, including 10 μg.
  • Recommended for biochemical assays, binding studies, and antibody characterization.
Catalog No. 50-001-95553