Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus / Rhesus macaque CTLA-4 Protein, Avitag™,His Tag
Supplier:  ACROBiosystems CT4C82E525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus macaque CTLA-4 are homologus.;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 15.9 kDa. The protein migrates as 20-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avitag,His Tag (Cat. No. CT4-C82E5) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avitag,His Tag (Cat. No. CT4-C82E5) with a linear range of 2-63 ng/mL (Routinely tested).
Catalog No. 50-195-7547